Learn More
4-Nitrodiphenylamine, 98+%
Product Code. p-7043425
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11380184 1 g 1g
11390184 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11380184 Supplier Thermo Scientific Alfa Aesar Supplier No. L00728.03
Add to basket
CAS: 836-30-6 | C12H10N2O2 | 214.22 g/mol
4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 836-30-6
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00007301
InChI Key XXYMSQQCBUKFHE-UHFFFAOYSA-N
Synonym 4-nitrodiphenylamine, p-nitrodiphenylamine, benzenamine, 4-nitro-n-phenyl, diphenylamine, 4-nitro, 4-nitrodifenylamin, 4-ndpa, p-nitrophenylphenylamine, 4-nitro-n-phenyl-aniline, 4-nitrophenyl phenylamine, 4-nitrodifenylamin czech
PubChem CID 13271
IUPAC Name 4-nitro-N-phenylaniline
SMILES [O-][N+](=O)C1=CC=C(NC2=CC=CC=C2)C=C1
Specifications
Melting Point 132°C to 135°C
Boiling Point 211°C (30 mmHg)
Flash Point 190°C (374°F)
Quantity 1 g
UN Number UN3077
Beilstein 2051910
Solubility Information Insoluble in water.
Formula Weight 214.22
Percent Purity ≥98%
Chemical Name or Material 4-Nitrodiphenylamine
compliance-icons
Product Identifier
  • 4-Nitrodiphenylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Nitrodiphenylamine