Learn More
4-Nitrobenzoyl Chloride, Sigma-Aldrich™
Product Code. 15886420
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886420 250 g 250g
15879330 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15886420 Supplier Sigma Aldrich Supplier No. 8067720250
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 122-04-3
Grade Research
Molecular Formula 4-(NO2)C6H4COCl
Molecular Weight (g/mol) 185.56
Spezifikation
Melting Point 75°C
Boiling Point 155°C/20 hPa
MDL Number MFCD00007345
Quantity 250 g
Synonym 4-Nitrobenzoyl chloride, 4-Nitrobenzoic acid chloride, 4-Nitrobenzoyl chloride