Learn More
4-Nitrobenzaldehyde, Sigma-Aldrich™
Product Code. 15839330
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839330 100 g 100g
15829330 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15839330 Supplier Sigma Aldrich Supplier No. 8067660100
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 555-16-8
Grade Research
Molecular Formula 4-(NO2)C6H4CHO
Molecular Weight (g/mol) 151.12
Specifications
Melting Point 101°C to 104°C
MDL Number MFCD00007346
Quantity 100 g
Synonym 4-Nitrobenzaldehyde, 4-Nitrobenzaldehyde