Learn More
Alfa Aesar™ 4-Nitro-1H-pyrazole, 97%
Product Code. 15433607
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433607 250 mg 250mg
11476150 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15433607 Supplier Alfa Aesar™ Supplier No. H27084.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 2075-46-9
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00159626
InChI Key XORHNJQEWQGXCN-UHFFFAOYSA-N
Synonym 4-nitropyrazole, 1h-pyrazole, 4-nitro, pyrazole, 4-nitro, 4-nitropyrrazole, 4-nitropyrazol, 4-nitro-pyrazole, 4-no2-pyrazole, 4-nitro-1h-pyrazol, 1h-pyrazole-4-nitro, pubchem14984
PubChem CID 16376
IUPAC Name 4-nitro-1H-pyrazole
SMILES [O-][N+](=O)C1=CNN=C1
Specifications
Melting Point 160°C to 162°C
Quantity 250 mg
Formula Weight 113.07
Percent Purity 97%
Chemical Name or Material 4-Nitro-1H-pyrazole