Learn More
4-(N-1-Dimethylaminonaphthalene-5-sulfonylamino)phenyl Isothiocyanate, TRC
Product Code. 30379828
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30379828 Supplier Toronto Research Chemicals Supplier No. TRCD46100010MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Chemical Name or Material 4-(N-1-Dimethylaminonaphthalene-5-sulfonylamino)phenyl Isoth
CAS 102417-94-7
Molecular Formula C19 H17 N3 O2 S2
Quantity 10 mg
InChI Formula InChI=1S/C19H17N3O2S2/c1-22(2)18-7-3-6-17-16(18)5-4-8-19(17)26(23,24)21-15-11-9-14(10-12-15)20-13-25/h3-12,21H,1-2H3
SMILES CN(C)c1cccc2c(cccc12)S(=O)(=O)Nc3ccc(cc3)N=C=S
IUPAC Name 5-(dimethylamino)-N-(4-isothiocyanatophenyl)naphthalene-1-sulfonamide
Molecular Weight (g/mol) 383.49
Formula Weight 383.0762
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.