Learn More
4-(Methylthiol)-1-(isothiocyanato)butane, TRC
Product Code. 30011523
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011523 50 mg 50mg
30009106 25 mg 25mg
30004048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011523 Supplier Toronto Research Chemicals Supplier No. TRCM33076350MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4430-36-8
Molecular Formula C6 H11 N S2
Molecular Weight (g/mol) 161.29
Synonym Erucin (4-(Methylthiol)-1-(Isothiocyanato)butane)
IUPAC Name 1-isothiocyanato-4-methylsulfanylbutane
SMILES CSCCCCN=C=S
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C6H11NS2/c1-9-5-3-2-4-7-6-8/h2-5H2,1H3
Formula Weight 161.0333
Chemical Name or Material Erucin