Learn More
4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanol, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanol |
| CAS | 1020719-61-2 |
| Molecular Formula | C10 2H3 H12 N3 O2 |
| Quantity | 10 mg |
| Synonym | 3-Pyridinemethanol, α-[3-(methyl-d3-nitrosoamino)propyl]-, α-[3-(Methyl-d3-nitrosoamino)propyl]-3-pyridinemethanol, (±)-NNAL-d3, NNAL-d3, 4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanol, 4-(Trideuteriomethylnitrosamino)-1-(3-pyridyl)-1-butanol, 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol-D3 (N-methyl-D3) |
| InChI Formula | InChI=1S/C10H15N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8,10,14H,3,5,7H2,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])N(CCCC(O)c1cccnc1)N=O |
| IUPAC Name | N-(4-hydroxy-4-pyridin-3-ylbutyl)-N-(trideuteriomethyl)nitrous amide |
| Molecular Weight (g/mol) | 212.26 |
| Formula Weight | 212.1353 |