Learn More
4-Methyl-2-pentanone, TRC
Product Code. 30481818
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481818 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30481818 Supplier Toronto Research Chemicals Supplier No. TRCM328025250G
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methyl-2-pentanone (MIBK)
CAS 108-10-1
Purity Grade Notes GC
Molecular Formula C6 H12 O
Quantity 250 g
Synonym 2-Pentanone, 4-methyl-, 4-Methyl-2-pentanone, 2-Methyl-4-pentanone, 2-Methylpropyl methyl ketone, 4-Methyl-2-oxopentane, Hexone, Isobutyl methyl ketone, Isopropylacetone, MIBK, MIK, Methyl 2-methylpropyl ketone, Methyl iso-butyl ketone, Methyl isobutyl ketone, NSC 5712, ZMD-D
InChI Formula InChI=1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3
SMILES CC(C)CC(=O)C
IUPAC Name 4-methylpentan-2-one
Molecular Weight (g/mol) 100.16
Formula Weight 100.0888
Percent Purity >95