Learn More
4-Methyl-2,3-dihydro-1H-inden-1-one, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-2,3-dihydro-1H-inden-1-one |
| CAS | 24644-78-8 |
| Molecular Formula | C10H10O |
| Quantity | 5 g |
| Synonym | 2,3-Dihydro-4-methyl-1H-inden-1-one, 4-Methyl-1-indanone |
| InChI Formula | InChI=1S/C10H10O/c1-7-3-2-4-9-8(7)5-6-10(9)11/h2-4H,5-6H2,1H3 |
| SMILES | Cc1cccc2C(=O)CCc12 |
| IUPAC Name | 4-methyl-2,3-dihydroinden-1-one |
| Molecular Weight (g/mol) | 146.19 |
| Formula Weight | 146.0732 |