Learn More
4'-Methoxyflavonol, MedChemExpress
Description
4'-Methoxyflavonol is a synthesized flavone/flavonol with 1, 2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) bilayers.
Specifications
Specifications
| Chemical Name or Material | 4'-Methoxyflavonol |
| CAS | 6889-78-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C16H12O4 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 6.4 mg/mL (23.86 mM; Need ultrasonic) |
| SMILES | O=C1C(O)=C(C2=CC=C(OC)C=C2)OC3=C1C=CC=C3 |
| Molecular Weight (g/mol) | 268.26 |
| Formula Weight | 268.26 |
| Show More |