Learn More
4-Methoxybenzoylacetonitrile, 98%
Product Code. 11428546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428546 5 g 5g
11438546 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11428546 Supplier Thermo Scientific Alfa Aesar Supplier No. A15973.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
CAS: 3672-47-7 | C10H9NO2 | 175.187 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3672-47-7
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00067889
InChI Key IKEPUFCALLUUBC-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-3-oxopropanenitrile, 4-methoxybenzoylacetonitrile, 4'-methoxybenzoylacetonitrile, 4-methoxybenzoyl acetonitrile, pubchem10102, 4-cyanoacetyl anisole, maybridge1_004582, acmc-1agd6, p-methoxybenzoylacetonitrile, toslab 6389
PubChem CID 233398
IUPAC Name 3-(4-methoxyphenyl)-3-oxopropanenitrile
SMILES COC1=CC=C(C=C1)C(=O)CC#N
Specifications
Melting Point 125°C to 130°C
Quantity 5 g
Beilstein 743909
Formula Weight 175.19
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzoylacetonitrile