Learn More
4-Methoxy Lornoxicam, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methoxy Lornoxicam |
| CAS | 2469039-23-2 |
| Molecular Formula | C14H12ClN3O4S2 |
| Quantity | 50 mg |
| Synonym | 6-Chloro-4-methoxy-2-methyl-N-(pyridin-2-yl)-2H-thieno[2,3-e][1,2]thiazine-3-carboxamide 1,1-dioxide, Lornoxicam Methyl Ether Impurity |
| InChI Formula | InChI=1S/C14H12ClN3O4S2/c1-18-11(14(19)17-10-5-3-4-6-16-10)12(22-2)13-8(24(18,20)21)7-9(15)23-13/h3-7H,1-2H3,(H,16,17,19) |
| SMILES | ClC(S1)=CC2=C1C(OC)=C(C(NC3=CC=CC=N3)=O)N(C)S2(=O)=O |
| Molecular Weight (g/mol) | 385.85 |