missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptobenzonitrile, TRC

Product Code. 30482727
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482727 1 g 1g
30467565 250 mg 250mg
30481644 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482727 Supplier Toronto Research Chemicals Supplier No. TRCM2526151G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36801-01-1
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.19
Synonym 4-Cyanobenzenethiol, 4-Cyanothiophenol, 4-Sulfanylbenzonitrile, p-Cyanobenzenethiol, p-Cyanobenzenethiol, p-Cyanothiophenol, p-Mercaptobenzonitrile
SMILES SC1=CC=C(C=C1)C#N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5NS/c8-5-6-1-3-7(9)4-2-6/h1-4,9H
Chemical Name or Material 4-Mercaptobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.