Learn More
4′-Isopropylacetophenone, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | 4′-Isopropylacetophenone |
| CAS | 645-13-6 |
| Boiling Range | 119°C to 120°C/13.3 hPa |
| Density | 0.970 g/cm³ at 20°C |
| Flash Point | > 110°C - (External MSDS) |
| Quantity | 10 mL |
| Synonym | 4′-Isopropylacetophenone, 4′-Isopropylacetophenone |
| Molecular Weight (g/mol) | 162.23 |
| Grade | Not Available |