Learn More
4-Isopropyl-3-methylaniline hydrochloride, 97%, Thermo Scientific™
Product Code. p-7024671
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408637 5 g 5g
11418637 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11408637 Supplier Thermo Scientific Alfa Aesar Supplier No. A19754.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Isopropyl-3-methylaniline hydrochloride is used to synthesis of fine chemical, organic intermediates.
Solubility
Slightly soluble in water (9.9 g/L)
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4534-11-6
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00156562
InChI Key VPOAQDSAPBSOLW-UHFFFAOYSA-N
Synonym 3-methyl-4-isopropylaniline hydrochloride, 4-isopropyl-3-methylaniline hydrochloride, 4-isopropyl-m-toluidinium chloride, benzenamine, 3-methyl-4-1-methylethyl-, hydrochloride, acmc-20a42e, 3-methyl-4-propan-2-ylaniline hydrochloride, 4-isopropyl-3-methylbenzenamine hydrochloride, 3-methyl-4-methylethyl phenylamine, chloride, 3-methyl-4-propan-2-yl aniline hydrochloride, benzenamine, 3-?methyl-?4-? 1-?methylethyl ?-?, hy
PubChem CID 2796270
IUPAC Name 3-methyl-4-propan-2-ylaniline;hydrochloride
SMILES CC1=C(C=CC(=C1)N)C(C)C.Cl
Specifications
Melting Point 222°C to 225°C
Quantity 5 g
Solubility Information Slightly soluble in water (9.9g/L)
Formula Weight 185.7
Percent Purity 97%
Chemical Name or Material 4-Isopropyl-3-methylaniline hydrochloride