missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Isopropoxy-2-methylbenzeneboronic acid, 98%

Product Code. 15459897
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459897 250 mg 250mg
15449897 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459897 Supplier Alfa Aesar™ Supplier No. H53132.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 871126-21-5
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.037
MDL Number MFCD07363788
InChI Key OEBXBUNQDNBWRP-UHFFFAOYSA-N
Synonym 4-isopropoxy-2-methylphenylboronic acid, 4-isopropoxy-2-methylphenyl boronic acid, 4-isopropoxy-2-methylbenzeneboronic acid, boronic acid,b-2-methyl-4-1-methylethoxy phenyl, 2-methyl-4-propan-2-yloxy phenyl boronic acid, acmc-209qf7, 4-isopropoxy-2-methylphenyl boronicacid, 4-isopropoxy-2-methyl-phenyl boronic acid, 2-methyl-4-propan-2-yl oxy phenyl boronic acid
PubChem CID 16218044
IUPAC Name (2-methyl-4-propan-2-yloxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OC(C)C)C)(O)O

Specifications

Melting Point 112°C to 116°C
Quantity 250 mg
Formula Weight 194.04
Percent Purity 98%
Chemical Name or Material 4-Isopropoxy-2-methylbenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.