Learn More
4-isocyanato-4-phenyltetrahydropyran, 97%, Thermo Scientific™
Product Code. 10235764
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10235764 250 mg Amber Glass Bottle 250mg
10472824 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10235764 Supplier Thermo Scientific Maybridge Supplier No. CC69106CB
 Restricted Item
Estimated Shipment: 05-01-2027
Chemical Identifiers
CAS 941717-02-8
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.24
MDL Number MFCD11506354
InChI Key HKKGHFQAYVPNAN-UHFFFAOYSA-N
Synonym 4-isocyanato-4-phenyltetrahydropyran, 4-isocyanato-4-phenyltetrahydro-2h-pyran, 4-phenyltetrahydro-2h-pyran-4-yl isocyanate, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-isocyanate
PubChem CID 33589534
IUPAC Name 4-isocyanato-4-phenyloxane
SMILES O=C=NC1(CCOCC1)C1=CC=CC=C1
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material 4-Isocyanato-4-phenyltetrahydropyran