missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobutyric acid, 98%, Thermo Scientific™

Product Code. 11954951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11954951 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11954951 Supplier Thermo Scientific Acros Supplier No. 438990010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7425-27-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 39°C
Color Orange to White
Boiling Point 109°C to 110°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C4H7IO2
Quantity 1 g
Synonym 4-iodobutyric acid, butanoic acid, 4-iodo, butanoicacid,4-iodo, gamma-iodo butyric acid, acmc-1bc28
InChI Key HALXOXQZONRCAB-UHFFFAOYSA-N
SMILES C(CC(=O)O)CI
IUPAC Name 4-iodobutanoic acid
Molecular Weight (g/mol) 214
PubChem CID 3354774
Formula Weight 214
Percent Purity 98%
Physical Form Low Melting Solid or Waxy Crystals
Chemical Name or Material 4-Iodobutyric acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodobutyric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.