missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-1H-pyrazole, 99%

Product Code. p-7037598
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15412838 1 g 1g
15422838 5 g 5g
15432838 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412838 Supplier Thermo Scientific Alfa Aesar Supplier No. H56369.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3469-69-0 | C3H3IN2 | 193.975 g/mol

4-Iodopyrazole was used in an indium-mediated synthesis of heterobiaryls. It undergoes iodination in the presence of iodine and ammonium hydroxide to yield 3,4-di-iodo- and 3,4,5-tri-iodo-pyrazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodopyrazole was used in an indium-mediated synthesis of heterobiaryls. It undergoes iodination in the presence of iodine and ammonium hydroxide to yield 3,4-di-iodo- and 3,4,5-tri-iodo-pyrazole.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3469-69-0
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.975
MDL Number MFCD00005244
InChI Key LLNQWPTUJJYTTE-UHFFFAOYSA-N
Synonym 4-iodopyrazole, 1h-pyrazole, 4-iodo, 4-iodo-pyrazole, pyrazole, 4-iodo, 4-iod-1h-pyrazol, 4-iodo pyrazole, sfthcqvilimkp@, zlchem 55, 4-iodo-1h-pyrazol, pubchem13295
PubChem CID 77022
ChEBI CHEBI:45260
IUPAC Name 4-iodo-1H-pyrazole
SMILES C1=C(C=NN1)I

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 193.97
Percent Purity 99%
Chemical Name or Material 4-Iodo-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.