missing translation for 'onlineSavingsMsg'
Learn More

(4-Imidazolyl)acetonitrile, 97%

Product Code. 15485777
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485777 5 g 5g
15475777 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485777 Supplier Thermo Scientific Alfa Aesar Supplier No. H32992.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18502-05-1 | C5H5N3 | 107.116 g/mol

(4-Imidazolyl)acetonitrile is used to produce 2-(1H-imidazol-4-yl)-3-phenyl-acrylonitrile at ambient temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(4-Imidazolyl)acetonitrile is used to produce 2-(1H-imidazol-4-yl)-3-phenyl-acrylonitrile at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18502-05-1
Molecular Formula C5H5N3
Molecular Weight (g/mol) 107.116
MDL Number MFCD00070306
InChI Key DQZBHUXBFNBJLX-UHFFFAOYSA-N
Synonym 2-1h-imidazol-4-yl acetonitrile, 1h-imidazole-5-acetonitrile, 1h-imidazole-4-acetonitrile, 2-1h-imidazol-5-yl acetonitrile, 1h-imidazol-4-yl-acetonitrile, 4 5-cyanomethylimidazole, 1h-imidazol-4-yl acetonitrile, 2-3h-imidazol-4-yl acetonitrile, 4-imidazolyl acetonitrile, 1h imidazole-4-acetonitrile
PubChem CID 571973
IUPAC Name 2-(1H-imidazol-5-yl)acetonitrile
SMILES C1=C(NC=N1)CC#N

Specifications

Melting Point 136°C to 138°C
Odor Odorless
Quantity 5 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 107.11
Percent Purity 97%
Chemical Name or Material (4-Imidazolyl)acetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.