missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymephenytoin, MedChemExpress

Product Code. 16492668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492668 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492668 Supplier Medchem Express Supplier No. HYW008772
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 4-Hydroxymephenytoin is a metabolism of an antiepileptic drug mephenytoin, which is used as a CYP2C19 substrate.

4-Hydroxymephenytoin is a metabolism of an antiepileptic drug mephenytoin, which is used as a CYP2C19 substrate.

Specifications

Chemical Name or Material 4-Hydroxymephenytoin
CAS 61837-65-8
Color White
Purity Grade Notes Research
Molecular Formula C12H14N2O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (426.89 mM; Need ultrasonic)
SMILES O=C1N(C)C(C(C2=CC=C(O)C=C2)(CC)N1)=O
Molecular Weight (g/mol) 234.25
Formula Weight 234.25
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.