missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl Isothiocyanate, TRC

Product Code. 30011630
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011630 500 mg 500mg
30010387 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011630 Supplier Toronto Research Chemicals Supplier No. TRCH829680500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2086-86-4
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
Synonym 4-Isothiocyanatomethyl-phenol, p-Hydroxybenzyl Ester Isothiocyanic Acid, 4-(Isothiocyanatomethyl)phenol, p-Hydroxybenzyl Isocyanate, p-Hydroxybenzyl Isothiocyanate;
IUPAC Name 4-(isothiocyanatomethyl)phenol
SMILES Oc1ccc(CN=C=S)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H7NOS/c10-8-3-1-7(2-4-8)5-9-6-11/h1-4,10H,5H2
Formula Weight 165.0248
Chemical Name or Material 4-Hydroxybenzyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.