Learn More
4-Hydroxybenzoylacrylic acid, 97%
Product Code. 11400348
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400348 5 g 5g
11490338 1 g 1g
11410348 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11400348 Supplier Thermo Scientific Alfa Aesar Supplier No. B24519.06
Request Bulk or Custom Format
CAS: 24849-48-7 | C10H7O4 | 191.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24849-48-7
Molecular Formula C10H7O4
Molecular Weight (g/mol) 191.16
MDL Number MFCD00460607
InChI Key LQCBGQDCRBYBMK-AATRIKPKSA-M
IUPAC Name (2E)-4-(4-hydroxyphenyl)-4-oxobut-2-enoate
SMILES OC1=CC=C(C=C1)C(=O)\C=C\C([O-])=O
Specifications
Melting Point 197°C to 198°C
Quantity 5 g
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzoylacrylic acid