Learn More
4-Hydroxybenzenesulfonic Acid Sodium Salt, TRC
Product Code. 30403935
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30403935 Supplier Toronto Research Chemicals Supplier No. TRCH8078101G
 Restricted Item
Estimated Shipment: 06-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Hydroxybenzenesulfonic Acid Sodium Salt
CAS 825-90-1
Molecular Formula C6H5NaO4S
Quantity 1 g
Synonym Sodium 4-Hydroxybenzenesulfonate, Sodium p-Hydroxybenzenesulfonate, Sodium p-Hydroxyphenylsulfonate, Sodium p-Phenolsulfonate, p-Hydroxybenzenesulfonic Acid Monosodium Salt, p-Hydroxybenzenesulfonic Acid Sodium Salt, p-Phenolsulfonic Acid Sodium Salt
InChI Formula InChI=1S/C6H6O4S.Na/c7-5-1-3-6(4-2-5)11(8,9)10;/h1-4,7H,(H,8,9,10);/q;+1/p-1
SMILES [Na+].Oc1ccc(cc1)S(=O)(=O)[O-]
IUPAC Name sodium;4-hydroxybenzenesulfonate
Molecular Weight (g/mol) 196.16
Formula Weight 195.9806