missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Propofol, TRC

Product Code. 30132698
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132698 10 mg 10mg
30007267 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132698 Supplier Toronto Research Chemicals Supplier No. TRCH95077010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1988-10-9
Molecular Formula C12 H18 O2
Molecular Weight (g/mol) 194.27
IUPAC Name 2,6-di(propan-2-yl)benzene-1,4-diol
SMILES CC(C)c1cc(O)cc(C(C)C)c1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H18O2/c1-7(2)10-5-9(13)6-11(8(3)4)12(10)14/h5-8,13-14H,1-4H3
Formula Weight 194.1307
Percent Purity >95%
Chemical Name or Material 4-Hydroxy Propofol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.