Learn More
4-Hydroxy Mexiletine Hydrochloride, TRC
Product Code. 30390963
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390963 25 mg -
30391121 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30390963 Supplier Toronto Research Chemicals Supplier No. TRCH94802025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123363-66-6
Molecular Formula C11 H17 N O2 . Cl H
Molecular Weight (g/mol) 231.72
Synonym Phenol, 4-(2-aminopropoxy)-3,5-dimethyl-, hydrochloride (1:1), Phenol, 4-(2-aminopropoxy)-3,5-dimethyl-, hydrochloride (9CI), KOE 2127CL, 4-Hydroxy Mexiletine hydrochloride, 4-Hydroxymexiletine hydrochloride, p-Hydroxymexiletine hydrochloride
IUPAC Name 4-(2-aminopropoxy)-3,5-dimethylphenol;hydrochloride
SMILES Cl.CC(N)COc1c(C)cc(O)cc1C
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C11H17NO2.ClH/c1-7-4-10(13)5-8(2)11(7)14-6-9(3)12;/h4-5,9,13H,6,12H2,1-3H3;1H
Formula Weight 231.1026
Chemical Name or Material 4-Hydroxy Mexiletine Hydrochloride