Learn More
4-Hydroxy-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Mikromol™
Product Code. 17911516
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911516 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17911516 Supplier Mikromol Supplier No. MM0084.240025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Hydroxy-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Hydroxy-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CAS 24683-22-5
Analyte or Component Names 4-Hydroxy-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
Shipping Condition Room Temperature
Molecular Formula C9 H8 N2 O4 S
Certifications/Compliance ISO 9001
Synonym 4-Hydroxy-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
InChI Formula InChI=1S/C9H8N2O4S/c10-9(13)7-8(12)5-3-1-2-4-6(5)16(14,15)11-7/h1-4,11-12H,(H2,10,13)
SMILES NC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1
IUPAC Name 4-hydroxy-1,1-dioxo-2H-1λ^{6},2-benzothiazine-3-carboxamide
Molecular Weight (g/mol) 240.24
Quantity 25 mg
Formula Weight 240.0205 g/mol