missing translation for 'onlineSavingsMsg'
Learn More

4-Hexyn-3-ol, 95%, Thermo Scientific™

Product Code. 11372237
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372237 25 g 25g
11362237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372237 Supplier Thermo Scientific Alfa Aesar Supplier No. L09727.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20739-59-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00041591
InChI Key KARLLBDFLHNKBO-UHFFFAOYNA-N
Synonym 4-hexyn-3-ol, ethyl 1-propynyl carbinol, 1-pentyn-3-ol, methyl, 4-hexyne-3-ol, acmc-1cpld
PubChem CID 140751
IUPAC Name hex-4-yn-3-ol
SMILES CCC(O)C#CC

Specifications

Density 0.894
Boiling Point 94°C to 95°C (105 mmHg)
Refractive Index 1.45
Quantity 25 g
UN Number UN1987
Beilstein 1739392
Formula Weight 98.15
Percent Purity 95%
Chemical Name or Material 4-Hexyn-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.