Learn More
4-Hexylresorcinol, Sigma-Aldrich™
Product Code. 15861091
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15861091 100 g 100g
15851091 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15861091 Supplier Sigma Aldrich Supplier No. 8206470100
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 136-77-6
Grade Research
Molecular Formula 4-[CH3(CH2)5]C6H3-1,3-(OH)2
Molecular Weight (g/mol) 194.27
Specifications
Melting Point 64°C to 67°C
Boiling Point 333°C to 335°C/1013 hPa
MDL Number MFCD00002284
Quantity 100 g
Synonym 4-Hexylresorcinol, 1,3-Dihydroxy-4-hexylbenzene, 1,3-Dihydroxy-4-hexylbenzene, 4-Hexylresorcinol