missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzhydrazide, 97%

Product Code. 15490378
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15490378

Brand: Thermo Scientific Alfa Aesar H53496.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 456-06-4 | C7H7FN2O | 154.144 g/mol

Used in the stnthesis of fluorinated poly (1, 3, 4-oxadiazole-ether-imide). N-(3,5-dinitrobenzoyl)-N?-(4-fluorobenzoyl)-hydrazine with 4-Fluorobenzhydrazide as solvent in NMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the stnthesis of fluorinated poly (1, 3, 4-oxadiazole-ether-imide). N-(3,5-dinitrobenzoyl)-N′-(4-fluorobenzoyl)-hydrazine with 4-Fluorobenzhydrazide as solvent in NMP.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-06-4
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.144
MDL Number MFCD00060562
InChI Key UIVXXFYJRYVRKJ-UHFFFAOYSA-N
Synonym 4-fluorobenzhydrazide, 4-fluorobenzoic hydrazide, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine, 4-fluorobenzoic acid hydrazide, hydrazine, p-fluorobenzoyl, 4-fluoro benzoyl hydrazine, benzoic acid, p-fluoro-, hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluoro-benzoic acid hydrazide
PubChem CID 9972
IUPAC Name 4-fluorobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)F

Specifications

Melting Point 160°C to 162°C
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzhydrazide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.