Learn More
4-Fluoro-3-[(trifluoromethyl)sulfonyl]benzenesulfonamide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Fluoro-3-[(trifluoromethyl)sulfonyl]benzenesulfonamide |
| CAS | 1027345-08-9 |
| Molecular Formula | C7H5F4NO4S2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C7H5F4NO4S2/c8-5-2-1-4(18(12,15)16)3-6(5)17(13,14)7(9,10)11/h1-3H,(H2,12,15,16) |
| SMILES | NS(=O)(=O)c1ccc(F)c(c1)S(=O)(=O)C(F)(F)F |
| Molecular Weight (g/mol) | 307.24 |
| Formula Weight | 306.9596 |