missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)aniline, 98%, Thermo Scientific™

Product Code. 11433787
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433787 25 g 25g
11423787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433787 Supplier Thermo Scientific Alfa Aesar Supplier No. A18250.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2357-47-3
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.118
MDL Number MFCD00007834
InChI Key PGFQDLOMDIBAPY-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl aniline, 5-amino-2-fluorobenzotrifluoride, alpha,alpha,alpha,4-tetrafluoro-m-toluidine, 3-trifluoromethyl-4-fluoroaniline, 4-fluoro-3-trifluoromethyl benzenamine, benzenamine, 4-fluoro-3-trifluoromethyl, unii-v5584x2ctl, 2-fluoro-5-aminobenzotrifluoride, 5-amino-2-fluoro trifluoromethyl benzene, 4-fluoro-3-trifluoromethyl phenylamine
PubChem CID 75377
IUPAC Name 4-fluoro-3-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1N)C(F)(F)F)F

Specifications

Density 1.393
Boiling Point 74°C to 75°C (10 mmHg)
Flash Point 91°C (195°F)
Odor Characteristic
Refractive Index 1.466
Quantity 25 g
UN Number UN2810
Beilstein 641587
Formula Weight 179.12
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.