accessibility menu, dialog, popup

UserName

CAS RN 2357-47-3

NH2FFFF

CAS RN 2357-47-3

IUPAC Name: 4-fluoro-3-(trifluoromethyl)aniline
Synonyms: 4-fluoro-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 179.12
Molecular Formula: C7H5F4N
InChi Key: PGFQDLOMDIBAPY-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C(=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results