missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-hydroxybenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15489287
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489287 250 mg 250mg
15479287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489287 Supplier Thermo Scientific Alfa Aesar Supplier No. H52550.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-3-hydroxybenzeneboronic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-74-2
Molecular Formula C6H6BFO3
Molecular Weight (g/mol) 155.919
MDL Number MFCD08689483
InChI Key GNCXNRGBOBWFSO-UHFFFAOYSA-N
Synonym 4-fluoro-3-hydroxyphenyl boronic acid, 4-fluoro-3-hydroxybenzeneboronic acid, 4-fluoro-3-hydroxy-phenyl boronic acid, 5-borono-2-fluorophenol, acmc-209r9z, 4-fluoro-3-hydroxy-phenylboronic acid, 4-fluoranyl-3-oxidanyl-phenyl boronic acid, boronic acid, b-4-fluoro-3-hydroxyphenyl
PubChem CID 44717470
IUPAC Name (4-fluoro-3-hydroxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)O)(O)O

Specifications

Melting Point 264°C to 266°C
Quantity 250 mg
Formula Weight 155.92
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-hydroxybenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.