missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-methylbenzyl bromide, 97%, Thermo Scientific™

Product Code. 11377009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377009 5 g 5g
11367009 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377009 Supplier Thermo Scientific Alfa Aesar Supplier No. H26849.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As pharmaceutical Intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862539-91-1
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.054
MDL Number MFCD03701059
InChI Key DAKPIAWEPVZDNG-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-fluoro-2-methylbenzene, 4-fluoro-2-methylbenzyl bromide, 4-fluoro-2-methylbenzylbromide, benzene, 1-bromomethyl-4-fluoro-2-methyl, 2-methyl-4-fluorobenzyl bromide, 4-fluoro-2-methyl-benzyl bromide, 1-bromomethyl-4-fluoro-2-methyl-benzene, benzene,1-bromomethyl-4-fluoro-2-methyl
PubChem CID 3732707
IUPAC Name 1-(bromomethyl)-4-fluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)F)CBr

Specifications

Refractive Index 1.5495
Quantity 5 g
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methylbenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.