missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-2-methoxyphenylacetic acid, 97%

Product Code. 15423467
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423467 250 mg 250mg
11421400 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423467 Supplier Alfa Aesar™ Supplier No. H26430.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886498-61-9
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.166
MDL Number MFCD06660193
InChI Key TUHNRCPWPMJXQN-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxyphenylacetic acid, 2-4-fluoro-2-methoxyphenyl acetic acid, 4-fluoro-2-methoxy phenylacetic acid, 4-fluoro-2-methoxybenzeneacetic acid, 4-fluoro-2-methoxyphenyl acetic acid
PubChem CID 17750773
IUPAC Name 2-(4-fluoro-2-methoxyphenyl)acetic acid
SMILES COC1=C(C=CC(=C1)F)CC(=O)O

Specifications

Melting Point 106°C to 110°C
Quantity 250 mg
Formula Weight 184.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-methoxyphenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.