missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynyltetrahydro-2H-pyran, TRC

Product Code. 30370375
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370375 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370375 Supplier Toronto Research Chemicals Supplier No. TRCE93262350MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1202245-65-5
Molecular Formula C7H10O
Quantity 50 mg
Synonym 4-Ethynyl-tetrahydro-2H-pyran
InChI Formula InChI=1S/C7H10O/c1-2-7-3-5-8-6-4-7/h1,7H,3-6H2
SMILES C#CC1CCOCC1
IUPAC Name 4-ethynyloxane
Molecular Weight (g/mol) 110.15
Formula Weight 110.0732
Chemical Name or Material 4-Ethynyltetrahydro-2H-pyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.