Learn More
4-Ethylguaiacol, TRC
Product Code. 30136837
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136837 10 g 10g
30008742 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136837 Supplier Toronto Research Chemicals Supplier No. TRCE67872010G
 Restricted Item
Estimated Shipment: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2785-89-9
Molecular Formula C9 H12 O2
Molecular Weight (g/mol) 152.19
Synonym Phenol, 4-ethyl-2-methoxy-, Guaiacol, 4-ethyl- (6CI), 4-Ethyl-2-methoxyphenol, 2-Methoxy-4-ethylphenol, 4-Ethylguaiacol, 4-Hydroxy-3-methoxyethylbenzene, 4-Hydroxy-3-methoxyphenylethane, Guaiacyl ethane, NSC 82313, p-Ethylguaiacol
IUPAC Name 4-ethyl-2-methoxyphenol
SMILES CCc1ccc(O)c(OC)c1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C9H12O2/c1-3-7-4-5-8(10)9(6-7)11-2/h4-6,10H,3H2,1-2H3
Formula Weight 152.0837
Chemical Name or Material 4-Ethylguaiacol