Learn More
4-Ethoxyphenylacetic Acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4919-33-9 |
| Molecular Formula | C10H12O3 |
| MDL Number | MFCD00004346 |
| Quantity | 5 g |
| SMILES | O=C(O)CC1=CC=C(OCC)C=C1 |
| Molecular Weight (g/mol) | 180.20 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Ethoxyphenylacetic Acid |