missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminoazobenzene-4'-Sulfonyl Chloride, 99.5%, MP Biomedicals™

Product Code. p-4774285 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11476702

Brand: MP Biomedicals 0215093183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromophoric label for amino acids, peptides, and proteins. Soluble in acetonitrile and DMF; slightly soluble in acetone

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56512-49-3
Molecular Formula C14H14ClN3O2S
Molecular Weight (g/mol) 323.80
MDL Number MFCD00007444
InChI Key VTVWTPGLLAELLI-UHFFFAOYSA-N
Synonym dabsyl chloride, 4-dimethylaminoazobenzene-4'-sulfonyl chloride, 4-dimethylamino azobenzene-4'-sulfonyl chloride, p-dimethylaminoazobenzene-4-sulfonyl chloride, p-p-dimethylamino phenyl azo benzenesulfonyl chloride, benzenesulfonyl chloride, 4-4-dimethylamino phenyl azo, benzenesulfonyl chloride, p-p-dimethylamino phenyl azo, 4-n,n-dimethylaminoazobenzene-4'-sulfonyl chloride, p-4-dimethylamino phenyl azo benzenesulphonyl chloride
PubChem CID 91660
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonyl chloride
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Color Red
Quantity 250 mg
Formula Weight 323.8
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminoazobenzene-4'-Sulfonyl Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.