Learn More
4-(Dimethylamino)benzaldehyde, Sigma-Aldrich™
Product Code. 15858910
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15858910 100 g 100g
15806340 250 g 250g
15816340 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15858910 Supplier Sigma Aldrich Supplier No. 8030570100
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-10-7
Grade Research
Molecular Formula 4-[(CH3)2N]C6H4CHO
Molecular Weight (g/mol) 149.19
Specifications
Melting Point 75°C
Density 1.22 g/cm3 at 20.1°C
MDL Number MFCD00003381
Quantity 100 g
Synonym 4-(Dimethylamino)benzaldehyde, p-Formyldimethylaniline, Ehrlich′s reagent, 4-(Dimethylamino)benzaldehyde