missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)butylamine, TRC

Product Code. 30384582
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30384582 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30384582

Brand: Toronto Research Chemicals TRCD443575100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Diethylamino)butylamine
CAS 27431-62-5
Molecular Formula C8H20N2
Quantity 100 mg
Synonym N,N-Diethyl-1,4-butanediamine, 4-(N,N-Diethylamino)butylamine, N,N-Diethyl-1,4-diaminobutane, N,N-Diethyltetramethylenediamine
InChI Formula InChI=1S/C8H20N2/c1-3-10(4-2)8-6-5-7-9/h3-9H2,1-2H3
SMILES CCN(CC)CCCCN
Molecular Weight (g/mol) 144.26
Formula Weight 144.1626
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.