Learn More
4-(Diethylamino)-2-butyn-1-ol, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 4-Diethylaminobut-2-yn-1-ol |
| CAS | 10575-25-4 |
| Molecular Formula | C8 H15 N O |
| Quantity | 500 mg |
| Synonym | 4-Diethylaminobut-2-yn-1-ol |
| InChI Formula | InChI=1S/C8H15NO/c1-3-9(4-2)7-5-6-8-10/h10H,3-4,7-8H2,1-2H3 |
| SMILES | CCN(CC)CC#CCO |
| IUPAC Name | 4-(diethylamino)but-2-yn-1-ol |
| Molecular Weight (g/mol) | 141.21 |
| Formula Weight | 141.1154 |