missing translation for 'onlineSavingsMsg'
Learn More

4-Decyne, 98%, Thermo Scientific™

Product Code. 11469007
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469007 25 g 25g
11459007 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469007 Supplier Thermo Scientific Alfa Aesar Supplier No. B20597.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Decyne is used in acid zinc plating.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2384-86-3
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
MDL Number MFCD00039978
InChI Key KVUNBQMRRNMQIZ-UHFFFAOYSA-N
Synonym 4-decyne, n-amyl n-propyl acetylene, acmc-1ch4b, n-pentyl n-propyl acetylene
PubChem CID 16944
IUPAC Name dec-4-yne
SMILES CCCCCC#CCCC

Specifications

Density 0.768
Boiling Point 75°C to 76°C (20 mmHg)
Flash Point 52°C (125°F)
Refractive Index 1.434
Quantity 25 g
UN Number UN3295
Beilstein 1698308
Solubility Information Insoluble in water.
Formula Weight 138.26
Percent Purity 98%
Chemical Name or Material 4-Decyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.