accessibility menu, dialog, popup

UserName

CAS RN 2384-86-3

H3CCH3

CAS RN 2384-86-3

IUPAC Name: dec-4-yne
Synonyms: dec-4-yne
Molecular Weight (g/mol): 138.25
Molecular Formula: C10H18
InChi Key: KVUNBQMRRNMQIZ-UHFFFAOYSA-N
SMILES: CCCCCC#CCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results