missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzaldehyde, Sigma-Aldrich™

Product Code. 15884421
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884421 25 g 25g
15874421 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15884421 Supplier Sigma-Aldrich Supplier No. 8185300025
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-07-7
Grade Research
Molecular Formula 4-(OHC)C6H4CN
Molecular Weight (g/mol) 131.13

Specifications

Melting Point 100°C to 103°C
Boiling Point 133°C/16 hPa
MDL Number MFCD00003376
Quantity 25 g
Synonym 4-Cyanobenzaldehyde, 4-Formylbenzonitrile, 4-Cyanobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.