Learn More
4-Cyano-3,5-difluorophenylboronic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Cyano-3,5-difluorophenylboronic acid |
| CAS | 861710-03-4 |
| Molecular Formula | C7 H4 B F2 N O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C7H4BF2NO2/c9-6-1-4(8(12)13)2-7(10)5(6)3-11/h1-2,12-13H |
| SMILES | OB(O)c1cc(F)c(C#N)c(F)c1 |
| IUPAC Name | (4-cyano-3,5-difluorophenyl)boronic acid |
| Molecular Weight (g/mol) | 182.92 |
| Formula Weight | 183.03 |