Learn More
4-cis-Fluoro-L-prolinamide hydrochloride, 98%
Product Code. 11372239
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372239 5 g 5g
15493637 250 mg 250mg
11362239 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11372239 Supplier Thermo Scientific Alfa Aesar Supplier No. H27235.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
CAS: 426844-23-7 | C5H10ClFN2O | 168.596 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 426844-23-7
Molecular Formula C5H10ClFN2O
Molecular Weight (g/mol) 168.596
MDL Number MFCD07368567
InChI Key YIEACUBSPQGCLW-MMALYQPHSA-N
Synonym 2s,4s-4-fluoropyrrolidine-2-carboxamide hydrochloride, 4-cis-fluoro-l-prolinamide hydrochloride, 2s,4s-4-fluoropyrrolidine-2-carboxamide hcl, cis-4-fluoro-l-prolinamide hydrochloride, 2s,4s-4-fluoro-2-pyrrolidinecarboxamide hydrochloride, 4s-4-fluoro-l-prolinamide-hydrogen chloride 1/1, 2s,4s-2-carbamoyl-4-fluoropyrrolidine hydrochloride salt, 2s,4s-4-fluoropyrrolidine-2-carboxamide monohydrochloride, 2s,4s-2-aminocarbonyl-4-fluoropyrrolidine hydrochloride
PubChem CID 45158917
IUPAC Name (2S,4S)-4-fluoropyrrolidine-2-carboxamide;hydrochloride
SMILES C1C(CNC1C(=O)N)F.Cl
Specifications
Melting Point 231°C to 236°C (decomposition)
Quantity 5 g
Optical Rotation −45 (c=1 in Water)
Formula Weight 168.59
Percent Purity 98%
Chemical Name or Material 4-cis-Fluoro-L-prolinamide hydrochloride