missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothioanisole, 98%

Product Code. 11418863
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418863 100 g 100g
11498853 5 g 5g
11408863 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418863 Supplier Thermo Scientific Alfa Aesar Supplier No. A11025.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 123-09-1 | C7H7ClS | 158.643 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-09-1
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00013643
InChI Key KIQQUVJOLVCZKG-UHFFFAOYSA-N
Synonym 4-chlorothioanisole, 4-chlorophenyl methyl sulfide, p-chlorothioanisole, 1-chloro-4-methylthio benzene, p-chlorophenyl methyl sulfide, methyl 4-chlorophenyl sulfide, benzene, 1-chloro-4-methylthio, p-chlophenyl methyl sulfide, 1-chloro-4-methylsulfanyl benzene, 4-chloro thioanisole
PubChem CID 31243
IUPAC Name 1-chloro-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C=C1)Cl

Specifications

Boiling Point 223°C
Flash Point 90°C (194°F)
Odor Unpleasant
Quantity 100 g
Beilstein 2041509
Formula Weight 158.65
Percent Purity 98%
Chemical Name or Material 4-Chlorothioanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.