Learn More
4-Chloroquinazoline, 95%, Thermo Scientific™
Product Code. 17760569 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17760569 5 g 5g
17750569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17760569 Supplier Thermo Scientific Acros Supplier No. 469400050
 Restricted Item
In Stock
Add to basket
Specifications
Alpha Vector CHLOROQUINAZOLINE
Chemical Name or Material 4-Chloroquinazoline
CAS 5190-68-1
Melting Point 100°C to 105°C
Color Brown to White
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C8 H5 Cl N2
MDL Number 00228682
Quantity 5 g
UN Number 2811
Molecular Weight (g/mol) 164.59
Percent Purity 0.95
Physical Form Crystals or Powder